Learn More
ACROBiosystems Cynomolgus DLL3 (27-488), His Tag (DL3-C52H9) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 488 (Accession # A0A2K5WSR1-1).
Catalog No. 502253343
Encompass_Preferred
Delta-like protein 3 (DLL3) is a transmembrane protein that belongs to the Delta/Serrate/Lag-2 (DSL) family of Notch ligands.May be required to divert neurons along a specific differentiation pathway. Plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. DLL3 protein is expressed on the surface of tumor cells but not in normal adult tissues.
This item is not returnable. View return policy
Catalog No. 50-225-3343 Supplier ACROBiosystems Supplier No. DL3C52H9100UG
Delta-like protein 3 (DLL3) is a transmembrane protein that belongs to the Delta/Serrate/Lag-2 (DSL) family of Notch ligands.May be required to divert neurons along a specific differentiation pathway. Plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. DLL3 protein is expressed on the surface of tumor cells but not in normal adult tissues.