Learn More
ACROBiosystems Biotinylated Cynomolgus DLL3 Protein, His,Avitag™
Catalog No. 502331346
Encompass_Preferred
Biotinylated Cynomolgus DLL3, His,Avitag (DL3-C82E9) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 488 (Accession # A0A2K5WSR1-1).
Catalog No. 50-233-1346 Supplier ACROBiosystems Supplier No. DL3C82E9200UG
Biotinylated Cynomolgus DLL3, His,Avitag (DL3-C82E9) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 488 (Accession # A0A2K5WSR1-1).