Learn More
ACROBiosystems Cynomolgus DLL3 Protein, His Tag
Catalog No. 502054900
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus DLL3, His Tag (DL3-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 490 (Accession # A0A2K5WSR4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4900 Supplier ACROBiosystems Supplier No. DL3C52H31MG
Recombinant Protein;1MG;Cynomolgus DLL3, His Tag (DL3-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 490 (Accession # A0A2K5WSR4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.