Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Nucleocapsid(D3L, R203K, G204R, S235F)-His Recombinant Protein
Supplier:  Sino Biological 40588V07E7100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Nucleocapsid Protein (YP_009724397.2) (Ser2-Ala419(D3L, R203K, G204R, S235F)) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-7242