Learn More
Sino Biological Cynomolgus DPP4 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 90180C01H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus DPP4 (F6VRB0) (Asp34-Pro766) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-1513