Learn More
Sino Biological Recombinant Cynomolgus Desmoglein-2 Protein (ECD, His Tag), HPLC-verified
Catalog No. 502992932
Encompass_Preferred
A DNA sequence encoding the Cynomolgus DSG2 (A0A2K5UGE1) (Ala50-Gly609) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2932 Supplier Sino Biological Supplier No. 91065C08H1
A DNA sequence encoding the Cynomolgus DSG2 (A0A2K5UGE1) (Ala50-Gly609) was expressed with a polyhistidine tag at the C-terminus.