Learn More
Sino Biological Cynomolgus EDAR Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90111C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus EDAR (F7GZI3) (Met1-Ala187) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2103