Learn More
Sino Biological Cynomolgus EDAR Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90111C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus EDAR (Met1-Ala187) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1119