Learn More
Sino Biological Recombinant Cynomolgus EPHA5 Protein (His Tag), HPLC-verified
Catalog No. 502993087
Encompass_Preferred
A DNA sequence encoding the Cynomolgus EPHA5 (XP_005555256.1) (Ser56-Pro573) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3087 Supplier Sino Biological Supplier No. 91049C08H100
A DNA sequence encoding the Cynomolgus EPHA5 (XP_005555256.1) (Ser56-Pro573) was expressed with a polyhistidine tag at the C-terminus.