Learn More
Sino Biological Recombinant Cynomolgus Her2/ERBB2 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 90295C49HB100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus ERBB2 (XP_001090430.1) (Met1-Thr652) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-9750