Learn More
Sino Biological Cynomolgus F9 Protein 500ug (Sino Biological)
Supplier:  Sino Biological 90996C08H500
Encompass_Preferred
A DNA sequence encoding the Cynomolgus F9 (A0A8J8XCV8) (Met1-Thr461) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1428