Learn More
Sino Biological Recombinant Cynomolgus CD32A/FCGR2A Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 90015C27HB100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus (Macaca fascicularis) CD32a (AAL92096.1) extracellular domain (Met 1-Ala 220) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-2739