Learn More
Sino Biological Recombinant Cynomolgus CD32A/FCGR2A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90015C08H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus (Macaca fascicularis) CD32a (AAL92096.1) extracellular domain (Met 1-Ile 211) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2740