Learn More
Sino Biological Cynomolgus FCGR2A Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90015C08H100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus (Macaca fascicularis) CD32a (AAL92096.1) extracellular domain (Met 1-Ile 211) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-0923