Learn More
Sino Biological Recombinant Anti-CD32B/Fcgr2b Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 90014R046P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus CD32b/FCGR2B/Fc gamma RIIB ( Catalog#90014-C08H; AAL92097.1; Met1-Pro217) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1964