Learn More
Sino Biological Recombinant Anti-CD32B/Fcgr2b Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 90014R046100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus CD32b / Fc gamma RIIB (Catalog#90014-C08H; AAL92097.1; Met1-Pro217).
Catalog No. 50-241-1971