Learn More
Sino Biological Recombinant Cynomolgus CD32B/Fcgr2b Protein (His Tag)
Supplier:  Sino Biological 90014C08H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus (Macaca fascicularis) CD32b (AAL92097.1) extracellular domain (Met 1-Pro 217) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2742