Learn More
Sino Biological Cynomolgus FCGRT & B2M Protein 20ug (Sino Biological)
Supplier:  Sino Biological CT031C08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Ser297) of cyno FCGRT (Q8SPV9) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the cyno B2M (Q8SPW0) (Met1-Met119) constructed the plasmid 2. The two plasmids were co-expressed and the FCGRT/B2M heterodimer was purified. The purified protein was biotinylated in vitro.
Catalog No. 50-237-2311