Learn More
Sino Biological Recombinant Cynomolgus FCGRT & B2M Heterodimer Protein (His Tag), HPLC-verified
Supplier:  Sino Biological CT031C08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 297) of cynomolgus FCGRT (Q8SPV9) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the cynomolgus B2M (Q8SPW0) (Met 1-Met 119) constructed the plasmid 2. The two plasmids were co-expressed and the FCGRT/B2M heterodimer was purified.
Catalog No. 50-255-2617