Learn More
ACROBiosystems Cynomolgus FCRL5 1mg
Supplier:  ACROBiosystems FC5C52H31MG
Encompass_Preferred
Cynomolgus FCRL5 Protein His Tag (FC5-C52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 16 - Gly 851 (Accession # XP_005541423.2).
Catalog No. 50-253-5085