Learn More
Sino Biological Cynomolgus FGF1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90062CNAE100
Encompass_Preferred
A DNA sequence encoding the cyno FGF1 (Phe16-Asp155) was expressed and purified with an initial Met.
Catalog No. 50-237-2237