Learn More
Sino Biological Recombinant Cynomolgus FGF-21 Protein (His Tag), HPLC-verified
Catalog No. 502992714
Encompass_Preferred
A DNA sequence encoding the Cynomolgus FGF21 (XP_005589868.1) (His29-Ser209) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-299-2714 Supplier Sino Biological Supplier No. 91068C07H1
A DNA sequence encoding the Cynomolgus FGF21 (XP_005589868.1) (His29-Ser209) was expressed, with a polyhistidine tag at the N-terminus.