Learn More
Sino Biological Cynomolgus FGL1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 90926C08B1
Encompass_Preferred
A DNA sequence encoding the cynomolgus FGL1 (XP_005562748.1) (Met37-Ile348) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2814