Learn More
ACROBiosystems Cynomolgus Flt-3 100ug
Catalog No. 502535064
Encompass_Preferred
Cynomolgus Flt-3 Protein His Tag (FL3-C52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Ser 541 (Accession # A0A2K5VQ48).
Catalog No. 50-253-5064 Supplier ACROBiosystems Supplier No. FL3C52H5100UG
Cynomolgus Flt-3 Protein His Tag (FL3-C52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Ser 541 (Accession # A0A2K5VQ48).