Learn More
Sino Biological Recombinant Cynomolgus Growth Hormone Receptor Protein (His Tag), HPLC-verified
Catalog No. 502992933
Encompass_Preferred
A DNA sequence encoding the Cynomolgus GHR (EHH62357.1) (Phe19-Tyr264) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2933 Supplier Sino Biological Supplier No. 91066C08H100
A DNA sequence encoding the Cynomolgus GHR (EHH62357.1) (Phe19-Tyr264) was expressed with a polyhistidine tag at the C-terminus.