Learn More
ACROBiosystems ACROBiosystems Cynomolgus GPA33 / A33 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems GP3C52H350UG
Encompass_Preferred
Recombinant Protein;50UG;Cynomolgus GPA33, His Tag (GP3-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # G7NU40-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 35-38 kDa and 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus GPA33, His Tag (Cat. No. GP3-C52H3) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GPA33 Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-210-6275