Learn More
Sino Biological Cynomolgus GPC3 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90956C08H100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus GPC3 (XP_005594665.1) (Met1-His559) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2305