Learn More
Sino Biological Recombinant Cynomolgus GPRC5D Protein (VLP, Full-length, Flag Tag)
Catalog No. 502993149
Encompass_Preferred
A DNA sequence encoding the Cynomolgus GPRC5D-VLP (Full Length) (A0A2K5W6I7) (Met1-Cys300) was fused with a signal peptide at the N-terminus and a flag tag at the C-terminus.
Catalog No. 50-299-3149 Supplier Sino Biological Supplier No. 91022C25H20
A DNA sequence encoding the Cynomolgus GPRC5D-VLP (Full Length) (A0A2K5W6I7) (Met1-Cys300) was fused with a signal peptide at the N-terminus and a flag tag at the C-terminus.