Learn More
Sino Biological Recombinant Cynomolgus Her2/ERBB2 Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological 90295C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus ERBB2 (Met1-Thr652) [similar to Macaca fascicularis ERBB2 (EHH58073.1)] was expressed with a polyhistidinetag at the C-terminus.
Catalog No. 50-224-6911