Learn More
PBL Assay Science VeriKine Cynomolgus Interferon Beta (IFN-Beta, IFNb) ELISA [1x96 well plate]
Catalog No. NC1644148 Shop All PBL Assay Science Products
Encompass
VeriKine Cynomolgus Interferon Beta ELISA is manufactured by PBL Assay Science (Catalog #46415-1) for research use only (RUO) and packaged as a 1x96-well plate. This assay is serum, plasma, and tissue culture media (TCM) compatible and has an assay range of 5.47-350 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.
Catalog No. NC1644148 Supplier PBL Assay Science Supplier No. 464151
VeriKine Cynomolgus Interferon Beta ELISA is manufactured by PBL Assay Science (Catalog #46415-1) for research use only (RUO) and packaged as a 1x96-well plate. This assay is serum, plasma, and tissue culture media (TCM) compatible and has an assay range of 5.47-350 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.