Learn More
Sino Biological Cynomolgus IFNA8 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 10119MM1520
Encompass_Preferred
A DNA sequence encoding the cynomolgus IFNA8(G7PS89) (Met1-Glu189) was expressed with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-237-4382