Learn More
ACROBiosystems Cynomolgus IgE Fc 1mg
Supplier:  ACROBiosystems IGEC52H31MG
Encompass_Preferred
Cynomolgus IgE Fc Protein His Tag (IGE-C52H3) is expressed from human 293 cells (HEK293). It contains AA Pro 111 - Lys 429 (Accession # G8F4W7).
Catalog No. 50-253-4627