Learn More
Sino Biological Anti-IGFBP-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90084T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus IGFBP3 (Catalog#90084-C07H; ; Gly28-Lys291). IGFBP3 specific IgG was purified by Cynomolgus IGFBP3 affinity chromatography.
Catalog No. 50-255-2722