Learn More
Sino Biological Anti-IGFBP4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 90080MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus IGFBP4 (Catalog#90080-C08H; Met1-Glu258). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.
Catalog No. 50-255-2729