Learn More
Sino Biological Anti-IGFBP4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90080T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus IGFBP4 (Catalog#90080-C08H; ; Met1-Glu258). IGFBP4 specific IgG was purified by Cynomolgus IGFBP4 affinity chromatography.
Catalog No. 50-255-2728