Learn More
Sino Biological Recombinant Cynomolgus IGFBP4 Protein (His Tag)
Supplier:  Sino Biological 90080C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus IGFBP4 (Met1-Glu258) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1088