Learn More
ACROBiosystems Cynomolgus IgG control 25mg
Catalog No. 502535139
Encompass_Preferred
Cynomolgus IgG1 Kappa Isotype Control (mAb) is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293) which combines the variable region of a cynomolgus monoclonal antibody with cynomolgus IgG1 constant domain. The cynomolgus monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a cynomolgus immunized with DNP.
Catalog No. 50-253-5139 Supplier ACROBiosystems Supplier No. DNPM55325MG
Cynomolgus IgG1 Kappa Isotype Control (mAb) is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293) which combines the variable region of a cynomolgus monoclonal antibody with cynomolgus IgG1 constant domain. The cynomolgus monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a cynomolgus immunized with DNP.