Learn More
ACROBiosystems Cynomolgus IL-1 Beta Protein, His Tag, Lyophilized, 20 μg
Catalog No. 502107066
Encompass_Preferred
Cynomolgus Interleukin-1 beta (IL-1β) is a cytokine protein involved in mediating inflammatory responses, including cell proliferation, differentiation, and apoptosis. This recombinant protein is expressed in E. coli and contains amino acids Ala 117 to Ser 268 of the Cynomolgus IL-1β sequence with an N-terminal polyhistidine tag.
  • Greater than 95% purity as determined by SDS-PAGE.
  • Calculated molecular weight of 19.2 kDa, migrating as 20 to 22 kDa under reducing conditions.
  • Formulated as a lyophilized powder from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose.
  • Endotoxin levels are less than 1.0 EU per μg.
  • Stable for 12 months in lyophilized state at -20°C to -70°C.
Catalog No. 50-210-7066 Supplier ACROBiosystems Supplier No. ILBC51H320UG
Cynomolgus Interleukin-1 beta (IL-1β) is a cytokine protein involved in mediating inflammatory responses, including cell proliferation, differentiation, and apoptosis. This recombinant protein is expressed in E. coli and contains amino acids Ala 117 to Ser 268 of the Cynomolgus IL-1β sequence with an N-terminal polyhistidine tag.
  • Greater than 95% purity as determined by SDS-PAGE.
  • Calculated molecular weight of 19.2 kDa, migrating as 20 to 22 kDa under reducing conditions.
  • Formulated as a lyophilized powder from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose.
  • Endotoxin levels are less than 1.0 EU per μg.
  • Stable for 12 months in lyophilized state at -20°C to -70°C.