Learn More
ACROBiosystems ACROBiosystems Cynomolgus IL-1 beta / IL-1F2 Protein, His Tag
Supplier:  ACROBiosystems ILBC51H31MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus IL-1 beta, His Tag (ILB-C51H3) is expressed from E.coli cells. It contains AA Ala 117 - Ser 268 (Accession # P79182-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.2 kDa. The protein migrates as 20-22 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-210-7064