Learn More
ACROBiosystems Cynomolgus IL-1 Rrp2 / IL-1 R6 Protein, His Tag
Supplier:  ACROBiosystems IL2C52H51MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus IL-1 Rrp2, His Tag (IL2-C52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 335 (Accession # EHH61702.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.9 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-205-5187