Learn More
ACROBiosystems Cynomolgus IL-10 Protein, His Tag
Catalog No. 502331211
Encompass_Preferred
Cynomolgus IL-10 Protein, His Tag (IL0-C4248) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Asn 178 (Accession # P79338).
Catalog No. 50-233-1211 Supplier ACROBiosystems Supplier No. IL0C424820UG
Cynomolgus IL-10 Protein, His Tag (IL0-C4248) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Asn 178 (Accession # P79338).