Learn More
ACROBiosystems Cynomolgus IL-17A / CTLA-8 Protein, His Tag
Catalog No. 502055269
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus IL-17A, His Tag (ILA-C5249) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (Accession # G7P4U9-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.0 kDa. The protein migrates as 18-19 kDa,24-25 kDa and 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus IL-17A, His Tag (Cat. No. ILA-C5249) at 2 μg/mL (100 μL/well) can bind Human IL-17 RA, Fc Tag (MALS verified) (Cat. No. ILA-H5257) with a linear range of 0.2-3 ng/mL (QC tested).;Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus IL-17A, His Tag (Cat. No. ILA-C5249) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human IL17A Antibody, Human IgG1 with a linear range of 0.1-0.8 ng/mL (Routinely tested).
Catalog No. 50-205-5269 Supplier ACROBiosystems Supplier No. ILAC52491MG
Recombinant Protein;1MG;Cynomolgus IL-17A, His Tag (ILA-C5249) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (Accession # G7P4U9-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.0 kDa. The protein migrates as 18-19 kDa,24-25 kDa and 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus IL-17A, His Tag (Cat. No. ILA-C5249) at 2 μg/mL (100 μL/well) can bind Human IL-17 RA, Fc Tag (MALS verified) (Cat. No. ILA-H5257) with a linear range of 0.2-3 ng/mL (QC tested).;Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus IL-17A, His Tag (Cat. No. ILA-C5249) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human IL17A Antibody, Human IgG1 with a linear range of 0.1-0.8 ng/mL (Routinely tested).