Learn More
ACROBiosystems Cynomolgus IL-2 R beta / CD122 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems ILBC52H91MG
Encompass_Preferred
Cynomolgus IL-2 R beta, His Tag (ILB-C52H9) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Thr 240 (Accession # Q38J85-1).
Catalog No. 50-233-1015