Learn More
Sino Biological Cynomolgus IL12RB1 Protein (His Tag) 100ug
Supplier:  Sino Biological 90980C08H100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus IL12RB1 (XP_015296347.1) (Met1-Asp545) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-222-1658