Learn More
Sino Biological Recombinant Cynomolgus IL-12 R beta 2/IL12RB2 Protein (ECD, mFc Tag), HPLC-verified
Supplier:  Sino Biological 90981C38H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus IL12RB2 (XP_005543136.1) (Lys24-Asn623) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-237-1631