Learn More
Sino Biological Recombinant Cynomolgus IL-13 R alpha 1/IL13RA1 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 90864C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus IL13RA1 (XP_005594500.1) (Met1-Thr341) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6565