Learn More
Sino Biological Recombinant Cynomolgus IL-1 beta Protein (His & AVI Tag), Avi-tag Biotinylated
Catalog No. 502992715
Encompass_Preferred
A DNA sequence encoding the Cynomolgus IL1B (P79182) (Ala117-Ser268) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2715 Supplier Sino Biological Supplier No. 91050C49HB20
A DNA sequence encoding the Cynomolgus IL1B (P79182) (Ala117-Ser268) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.