Learn More
Sino Biological Anti-IL-1 beta Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90010RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus IL-1 beta (Catalog#90010-CNAE; NP_001036221.1; Ala117-Ser269). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2259