Learn More
Sino Biological Anti-IL1R1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 90279MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus IL-1R1 / CD121a (Catalog#90279-C08H; EHH61701.1; Met1-Thr332).
Catalog No. 50-241-2284