Learn More
Sino Biological Anti-IL1R1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90279T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus IL-1R1 / CD121a (Catalog#90279-C08H; EHH61701.1; Met1-Thr332). IL-1R1 / CD121a specific IgG was purified by Cynomolgus IL-1R1 / CD121a affinity chromatography.
Catalog No. 50-241-0349